3'-(Trifluoromethyl)biphenyl-3-carboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4840-1G
CA-4840-5G
COBBCA-4840-1GEA
0
GBP
COBBCA-4840-1G
COBBCA-4840-5G
3'-(Trifluoromethyl)biphenyl-3-carboxylic acid
3'-(Trifluoromethyl)biphenyl-3-carboxylic acid
Formula:
C₁₄H₉F₃O₂ MW: 266.22 g/mol |
MDL Number:
MFCD03424603 CAS Number: 168619-05-4 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...