4-Amino-2-(trifluoromethyl)benzonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Cyano-3-trifluoromethylaniline

M-Clarity™ quality level = MQ100

631752-1G 631752-5G
SIAL631752-1GEA 38.8 GBP
SIAL631752-1G SIAL631752-5G
4-Amino-2-(trifluoromethyl)benzonitrile, Sigma-Aldrich®
4-Amino-2-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
MDL Number: MFCD00042155
CAS Number: 654-70-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR