4-Amino-2-(trifluoromethyl)benzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Cyano-3-trifluoromethylaniline

CFF185.2.5 CFF185.10 CFF185.25 003971-25G 003971-100G
CHMPCFF185.2.5EA 0 GBP
CHMPCFF185.2.5 CHMPCFF185.10 CHMPCFF185.25 FLUO003971-25G FLUO003971-100G
4-Amino-2-(trifluoromethyl)benzonitrile
4-Amino-2-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
MDL Number: MFCD00042155
CAS Number: 654-70-6
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR