1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

00738-50G-F 00738-5G-F
SIAL00738-50G-FEA 875 GBP
SIAL00738-50G-F SIAL00738-5G-F
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
Formula: C₇H₁₁F₃N₂O₃S
MDL Number: MFCD00210008
CAS Number: 145022-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR