5-Fluorogramine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(5-Fluoro-1H-indol-3-yl)-N,N-dimethylmethanamine

010186-1G 010186-5G
FLUO010186-1GEA 0 GBP
FLUO010186-1G FLUO010186-5G
5-Fluorogramine
5-Fluorogramine
Formula: C₁₁H₁₃FN₂
MW: 192.24 g/mol
MDL Number: MFCD00022731
CAS Number: 343-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR