1-Boc-4-nitroindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0151-5G IN-0151-1G
COBBIN-0151-5GEA 0 GBP
COBBIN-0151-5G COBBIN-0151-1G
1-Boc-4-nitroindole
1-Boc-4-nitroindole
Formula: C₁₃H₁₄N₂O₄
MDL Number: MFCD08689543
CAS Number: 913836-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR