Methyl indole-7-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 1H-indole-7-carboxylate

IN-0020-5G IN-0020-25G IN-0020-1G
COBBIN-0020-5GEA 0 GBP
COBBIN-0020-5G COBBIN-0020-25G COBBIN-0020-1G
Methyl indole-7-carboxylate
Methyl indole-7-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00211064
CAS Number: 93247-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR