Methyl indole-3-carboxylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003455-25G 003455-100G 003455.500G
FLUO003455-25GEA 0 GBP
FLUO003455-25G FLUO003455-100G FLUO003455.500G
Methyl indole-3-carboxylate
Methyl indole-3-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00189407
CAS Number: 942-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR