1-Boc-7-bromoindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 7-Bromo-1H-indole, N-BOC protected

IN-0163-5G IN-0163-1G
COBBIN-0163-5GEA 0 GBP
COBBIN-0163-5G COBBIN-0163-1G
1-Boc-7-bromoindole
1-Boc-7-bromoindole
Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
MDL Number: MFCD09475841
CAS Number: 868561-17-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR