1-Boc-5-bromoindole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-Boc-5-bromoindole, 5-Bromo-1-tert-butoxycarbonylindole, tert-Butyl 5-bromo-1H-indole-1-carboxylate
MQ100
557749-25G-A
557749-5G-A
SIAL557749-25G-AEA
499
GBP
SIAL557749-25G-A
SIAL557749-5G-A
1-Boc-5-bromoindole, Sigma-Aldrich®
1-Boc-5-bromoindole
Formula:
C₁₃H₁₄BrNO₂ MW: 296.16 g/mol |
MDL Number:
MFCD02090067 CAS Number: 182344-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...