1-Boc-4-bromoindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0152-5G IN-0152-1G
COBBIN-0152-5GEA 0 GBP
COBBIN-0152-5G COBBIN-0152-1G
1-Boc-4-bromoindole
1-Boc-4-bromoindole
Formula: C₁₃H₁₄BrNO₂
MDL Number: MFCD04117874
CAS Number: 676448-17-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR