5-(Trifluoromethoxy)oxindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0148-5G IN-0148-1G
COBBIN-0148-5GEA 0 GBP
COBBIN-0148-5G COBBIN-0148-1G
5-(Trifluoromethoxy)oxindole
5-(Trifluoromethoxy)oxindole
Formula: C₉H₆F₃NO₂
MDL Number: MFCD09027275
CAS Number: 403853-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR