5-Bromooxindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0071-5G IN-0071-1G
COBBIN-0071-5GEA 0 GBP
COBBIN-0071-5G COBBIN-0071-1G
5-Bromooxindole
5-Bromooxindole
Formula: C₈H₆BrNO
MDL Number: MFCD00456998
CAS Number: 20870-78-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR