1-Boc-3-piperidone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011528-5G 011528-100G 011528-25G
FLUO011528-5GEA 0 GBP
FLUO011528-5G FLUO011528-100G FLUO011528-25G
1-Boc-3-piperidone
1-Boc-3-piperidone
Formula: C₁₀H₁₇NO₃
MDL Number: MFCD01631193
CAS Number: 98977-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR