Geraniol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

M-Clarity™ quality level = MQ100

163333-100G 163333-25G
SIAL163333-100GEA 94.8 GBP
SIAL163333-100G SIAL163333-25G
Geraniol, Sigma-Aldrich®
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR