1-Boc-4-chloroindole-2-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chloro-N-(Boc)-indole-2-boronic acid, (1-(tert-Butoxycarbonyl)-4-chloro-1H-indol-2-yl)boronic acid

BB-3493-5G BB-3493-25G BB-3493-1G
COBBBB-3493-5GEA 0 GBP
COBBBB-3493-5G COBBBB-3493-25G COBBBB-3493-1G
1-Boc-4-chloroindole-2-boronic acid
1-Boc-4-chloroindole-2-boronic acid
Formula: C₁₃H₁₅BClNO₄
MW: 295.53 g/mol
MDL Number: MFCD04114582
CAS Number: 475102-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR