3-Bromo-2-nitroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1330-5G AN-1330-25G AN-1330-1G
COBBAN-1330-5GEA 0 GBP
COBBAN-1330-5G COBBAN-1330-25G COBBAN-1330-1G
3-Bromo-2-nitroaniline
3-Bromo-2-nitroaniline
Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD08063248
CAS Number: 7138-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR