2,4-Dibromo-3,5-difluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024169-25G 024169-5G
FLUO024169-25GEA 0 GBP
FLUO024169-25G FLUO024169-5G
2,4-Dibromo-3,5-difluoroaniline
2,4-Dibromo-3,5-difluoroaniline
Formula: C₆H₃Br₂F₂N
MW: 286.9 g/mol
MDL Number: MFCD06797746
CAS Number: 883549-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR