2-Amino-4,6-difluorobenzothiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,6-Difluorobenzothiazol-2-ylamine

CFF767.1 005535-1G 005535-2.5G
CHMPCFF767.1EA 0 GBP
CHMPCFF767.1 FLUO005535-1G FLUO005535-2.5G
2-Amino-4,6-difluorobenzothiazole
2-Amino-4,6-difluorobenzothiazole
Formula: C₇H₄F₂N₂S
MW: 186.18 g/mol
MDL Number: MFCD00276099
CAS Number: 119256-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR