6-Fluorosalicylaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-6-hydroxybenzaldehyde

013018-1G 013018-250MG 013018-5G
FLUO013018-1GEA 0 GBP
FLUO013018-1G FLUO013018-250MG FLUO013018-5G
6-Fluorosalicylaldehyde
6-Fluorosalicylaldehyde
Formula: C₇H₅FO₂
MDL Number: MFCD01090996
CAS Number: 38226-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR