2-Bromo-5-(trifluoromethoxy)aniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7467-5G
OS-7467-25G
OS-7467-1G
COBBOS-7467-5GEA
0
GBP
COBBOS-7467-5G
COBBOS-7467-25G
COBBOS-7467-1G
2-Bromo-5-(trifluoromethoxy)aniline
2-Bromo-5-(trifluoromethoxy)aniline
Formula:
C₇H₅BrF₃NO |
MDL Number:
MFCD08458013 CAS Number: 887267-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...