2,6-Dibromo-4-(trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004677-1G 004677-5G 004677-25G 004677-100G
FLUO004677-1GEA 0 GBP
FLUO004677-1G FLUO004677-5G FLUO004677-25G FLUO004677-100G
2,6-Dibromo-4-(trifluoromethoxy)aniline
2,6-Dibromo-4-(trifluoromethoxy)aniline
Formula: C₇H₄Br₂F₃NO
MDL Number: MFCD00153113
CAS Number: 88149-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR