Erbium triacetate hydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Erbium(III) acetate hydrate

M-Clarity™ quality level = MQ100

325570-10G 325570-50G
SIAL325570-10GEA 126 GBP
SIAL325570-10G SIAL325570-50G
Erbium triacetate hydrate, Sigma-Aldrich®
Erbium triacetate hydrate
Formula: C₆H₉ErO₆·H₂O
CAS Number: 304675-52-3
EINECS: 247-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR