2-Chloro-6-fluoro-m-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-6-fluoro-3-methylbenzoic acid

005646-5G 005646-10G
FLUO005646-5GEA 0 GBP
FLUO005646-5G FLUO005646-10G
2-Chloro-6-fluoro-m-toluic acid
2-Chloro-6-fluoro-m-toluic acid
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
MDL Number: MFCD01631359
CAS Number: 32890-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR