2-(Dimethylamino)benzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF216.1
CFF216.2.5
017275-1G
017275-2.5G
CHMPCFF216.1EA
0
GBP
CHMPCFF216.1
CHMPCFF216.2.5
FLUO017275-1G
FLUO017275-2.5G
2-(Dimethylamino)benzaldehyde
2-(Dimethylamino)benzaldehyde
Formula:
C₉H₁₁NO MW: 149.19 g/mol |
MDL Number:
MFCD03407377 CAS Number: 579-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...