1,2,3,4-Tetrahydroquinoline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

T15504-100G T15504-500G T15504-5G
SIALT15504-100GEA 89.3 GBP
SIALT15504-100G SIALT15504-500G SIALT15504-5G
1,2,3,4-Tetrahydroquinoline, Sigma-Aldrich®
1,2,3,4-Tetrahydroquinoline
Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00006693
CAS Number: 635-46-1
EINECS: 211-237-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR