D-allo-Threonine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-D-Allo-Thr-OH, (2R,3R)-2-Amino-3-hydroxybutanoic acid

4008353.0250 4008353.1000
BACE4008353.0250EA 0 GBP
BACE4008353.0250 BACE4008353.1000
D-allo-Threonine
D-allo-Threonine
Formula: C₄H₉NO₃
MDL Number: MFCD00004526
CAS Number: 24830-94-2
EINECS: 246-488-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR