(S)-(+)-2-Aminobutyric acid

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid

4030313.0005 4030313.0025
BACE4030313.0005EA 59.55 GBP
BACE4030313.0005 BACE4030313.0025
(S)-(+)-2-Aminobutyric acid
(S)-(+)-2-Aminobutyric acid
Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00064415
CAS Number: 1492-24-6
EINECS: 216-083-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR