2,6-Dibromo-4-tert-butylaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023856-25G 023856-5G
FLUO023856-25GEA 0 GBP
FLUO023856-25G FLUO023856-5G
2,6-Dibromo-4-tert-butylaniline
2,6-Dibromo-4-tert-butylaniline
Formula: C₁₀H₁₃Br₂N
MDL Number: MFCD06657952
CAS Number: 10546-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR