Methylmalonic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methylmalonic acid
M-Clarity™ quality level = MQ200
M54058-5G
M54058-25G
SIALM54058-5GEA
72.8
GBP
SIALM54058-5G
SIALM54058-25G
Methylmalonic acid, Sigma-Aldrich®
Methylmalonic acid
Formula:
C₄H₆O₄ MW: 118.09 g/mol |
MDL Number:
MFCD00002656 CAS Number: 516-05-2 EINECS: 208-219-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...