1-Undecanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane

U1001-100G U1001-500G
SIALU1001-100GEA 61.3 GBP
SIALU1001-100G SIALU1001-500G
1-Undecanol, Sigma-Aldrich®
1-Undecanol
Formula: C₁₁H₂₄O
MW: 172.31 g/mol
Boiling Pt: 243
Melting Pt: 13…15 °C
Density: 0.83 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004751
CAS Number: 112-42-5
EINECS: 203-970-5
UN: 3082
ADR: 9 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR