Octylamine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Aminooctane, n-Octylamine, 1-Octylamine
O5802-100G
O5802-500G
O5802-5G
SIALO5802-100GEA
42
GBP
SIALO5802-100G
SIALO5802-500G
SIALO5802-5G
Octylamine, Sigma-Aldrich®
Octylamine
Formula:
H₃C(CH₂)₆CH₂NH₂ MW: 129.25 g/mol Boiling Pt: 180 Melting Pt: –1 °C Density: 0.7812 g/cm³ (20 °C) Flash Pt: 60 Storage Temperature: Ambient |
MDL Number:
MFCD00008247 CAS Number: 111-86-4 EINECS: 203-916-0 UN: 2734 ADR: 8 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...