5-Bromo-2-iodobenzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-iodobenzotrifluoride, 1-Bromo-4-iodo-3-(trifluoromethyl)benzene

13247.06 013247-25G 013247-5G
FLUO13247.06EA 0 GBP
FLUO13247.06 FLUO013247-25G FLUO013247-5G
5-Bromo-2-iodobenzotrifluoride
5-Bromo-2-iodobenzotrifluoride
Formula: C₇H₃BrF₃I
MDL Number: MFCD00672935
CAS Number: 364-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR