4-Fluoro-3-(trifluoromethyl)benzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Cyano-2-fluorobenzotrifluoride

AN-1049-5G AN-1049-25G AN-1049-100G
COBBAN-1049-5GEA 0 GBP
COBBAN-1049-5G COBBAN-1049-25G COBBAN-1049-100G
4-Fluoro-3-(trifluoromethyl)benzonitrile
4-Fluoro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
Boiling Pt: 193…195 °C (1013 hPa)
Melting Pt: 67…68 °C
MDL Number: MFCD00061284
CAS Number: 67515-59-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR