2-Thiouracil, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Hydroxypyrimidine-2(1H)-thione
T7750-1KG
T7750-100G
T7750-250G
T7750-25G
SIALT7750-1KGEA
970
GBP
SIALT7750-1KG
SIALT7750-100G
SIALT7750-250G
SIALT7750-25G
2-Thiouracil, Sigma-Aldrich®
2-Thiouracil
Formula:
C₄H₄N₂OS Melting Pt: 340 °C |
MDL Number:
MFCD00006039 CAS Number: 141-90-2 EINECS: 205-508-8 Merck Index: 12,09504 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...