2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine

PN-6056-1G PN-6056-5G PN-6056-250M
COBBPN-6056-1GEA 0 GBP
COBBPN-6056-1G COBBPN-6056-5G COBBPN-6056-250M
2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester
2-Chloro-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₄BClF₃NO₂
MDL Number: MFCD13195760
CAS Number: 1218790-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR