2-Fluoro-6-(trifluoromethyl)benzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Fluoro-6-(trifluoromethyl)phenyl)methanol
006412-1G
006412-5G
006412-10G
FLUO006412-1GEA
0
GBP
FLUO006412-1G
FLUO006412-5G
FLUO006412-10G
2-Fluoro-6-(trifluoromethyl)benzyl alcohol
2-Fluoro-6-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₆F₄O MW: 194.13 g/mol |
MDL Number:
MFCD00061168 CAS Number: 152211-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...