Trichloroacetonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
T53805-100G
T53805-500G
T53805-5G
SIALT53805-100GEA
99.3
GBP
SIALT53805-100G
SIALT53805-500G
SIALT53805-5G
Trichloroacetonitrile, Sigma-Aldrich®
Trichloroacetonitrile
Formula:
C₂Cl₃N MW: 144.39 g/mol Boiling Pt: 85.7 Melting Pt: –44 °C Density: 1.44 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001842 CAS Number: 545-06-2 EINECS: 208-885-7 UN: 3276 ADR: 6.1 III Merck Index: 12,09757 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...