3,4,5-Trimethoxyphenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4,4,5,5-Tetramethyl-2-(3,4,5-trimethoxyphenyl)-1,3,2-dioxaborolane
PN-8323-5G
PN-8323-1G
COBBPN-8323-5GEA
0
GBP
COBBPN-8323-5G
COBBPN-8323-1G
3,4,5-Trimethoxyphenylboronic acid pinacol ester
3,4,5-Trimethoxyphenylboronic acid pinacol ester
Formula:
C₁₅H₂₃BO₅ MW: 294.1511 g/mol |
MDL Number:
MFCD12405526 CAS Number: 214360-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...