3,4,5-Trifluorophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-2031-5G PN-2031-1G
COBBPN-2031-5GEA 0 GBP
COBBPN-2031-5G COBBPN-2031-1G
3,4,5-Trifluorophenylboronic acid pinacol ester
3,4,5-Trifluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₄BF₃O₂
MDL Number: MFCD05663885
CAS Number: 827614-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR