4-Pentynoic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
232211-1G
232211-5G
SIAL232211-1GEA
48.2
GBP
SIAL232211-1G
SIAL232211-5G
4-Pentynoic acid, Sigma-Aldrich®
4-Pentynoic acid
Formula:
C₅H₆O₂ MW: 98.1 g/mol Boiling Pt: 199 °C (1013 hPa) Melting Pt: 57 °C |
MDL Number:
MFCD00004407 CAS Number: 6089-09-4 EINECS: 228-028-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...