2-Aminopurine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

A3509-100MG A3509-250MG A3509-500MG A3509-1G
SIALA3509-100MGEA 160 GBP
SIALA3509-100MG SIALA3509-250MG SIALA3509-500MG SIALA3509-1G
2-Aminopurine, Sigma-Aldrich®
2-Aminopurine
Formula: C₅H₅N₅
Melting Pt: 277…278 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005566
CAS Number: 452-06-2
EINECS: 207-197-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR