Hydroquinone monomethyl ether, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole
M-Clarity™ quality level = MQ200
54050-100G
54050-500G
SIAL54050-100GEA
22.9
GBP
SIAL54050-100G
SIAL54050-500G
Hydroquinone monomethyl ether, Sigma-Aldrich®
Hydroquinone monomethyl ether
Formula:
C₇H₈O₂ MW: 124.14 g/mol Boiling Pt: 243 °C (1013 hPa) Melting Pt: 55…57 °C Density: 1.55 g/cm³ (20 °C) Flash Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002332 CAS Number: 150-76-5 EINECS: 205-769-8 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...