N-Ethylaniline ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N-Ethyl-N-phenylamine
A15060.0F
A15060.AE
A15060.AP
A15060.0FEA
75.9
GBP
A15060.0F
A15060.AE
A15060.AP
N-Ethylaniline ≥97%
N-Ethylaniline
Purity: 97%
CAS No.: 103-69-5
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 85°(185°F)
Refractive Index: 1.5540
Odour: Odourless
Melting Point: -64°
Boiling Point: 204-205°
Beilstein Reference: 507468
Merck Reference: 14,3764
UN No.: 2272
MDL No.: MFCD00009025
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 204…205 °C Melting Pt: –64 °C Density: 0.961 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009025 CAS Number: 103-69-5 EINECS: 203-135-5 UN: 2272 ADR: 6.1,III Merck Index: 14,3764 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...