3,4,5-Trimethoxyaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
008860-5G
008860-100G
008860-25G
FLUO008860-5GEA
0
GBP
FLUO008860-5G
FLUO008860-100G
FLUO008860-25G
3,4,5-Trimethoxyaniline
3,4,5-Trimethoxyaniline
Formula:
C₉H₁₃NO₃ Boiling Pt: 293 °C (1013 hPa) Melting Pt: 110…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008393 CAS Number: 24313-88-0 EINECS: 246-154-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...