Incubators, Premier range
Supplier: GENLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRI/30 PRI/100 PRI/125 PRI/150 PRI/200 PRI/75 PRI/50
390-0484EA 1430 GBP
390-0484 390-0487 390-0488 390-0489 390-0490 390-0486 390-0485
Incubators, Premier range
Incubators Standard Incubators
Genlab Premier incubators (PRI) are a range of fully functional, highly efficient units to suit most biological analysis, research and general laboratory applications. The exterior is constructed from sheet steel finished in an easy clean, powder coated paint and the interior chamber is made from stainless steel. The PRI incubators have a solid steel outer door with a glass inner door.
  • PID microprocessor controller with dual display of set point and actual temperature, which are auto-tuned for each individual unit to optimise the heat up, overshoot and
More Product Information