4-Iodoaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
005423-25G
005423-100G
FLUO005423-25GEA
0
GBP
FLUO005423-25G
FLUO005423-100G
4-Iodoaniline
4-Iodoaniline
Formula:
C₆H₆IN Boiling Pt: 262 °C (1013 hPa) Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007848 CAS Number: 540-37-4 EINECS: 208-743-4 Merck Index: 13,05049 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...