2-Iodoaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001038-25G
001038-10G
001038-50G
FLUO001038-25GEA
0
GBP
FLUO001038-25G
FLUO001038-10G
FLUO001038-50G
2-Iodoaniline
Formula:
C₆H₆IN Boiling Pt: 263 °C (1013 hPa) Melting Pt: 54…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007680 CAS Number: 615-43-0 EINECS: 210-426-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...