5-Iodovanillin

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde

002992-10G 002992-50G
FLUO002992-10GEA 0 GBP
FLUO002992-10G FLUO002992-50G
5-Iodovanillin
5-Iodovanillin
Formula: C₈H₇IO₃
MW: 278.05 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 186…187 °C
MDL Number: MFCD00006941
CAS Number: 5438-36-8
EINECS: 226-617-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR