Iodobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Phenyliodide
003461-500G
003461-100G
FLUO003461-500GEA
0
GBP
FLUO003461-500G
FLUO003461-100G
Iodobenzene
Iodobenzene
Formula:
C₆H₅I MW: 204.01 g/mol Boiling Pt: 188 °C (1013 hPa) Melting Pt: –30 °C Density: 1.838 g/cm³ (20 °C) Flash Pt: 74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001029 CAS Number: 591-50-4 EINECS: 209-719-6 Merck Index: 13,05051 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...